siRNA Details
Gene Symbol: Gsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004080.1 | NP_001004080.1 | 4 | 633 | ||
XM_006234039.2 | XP_006234101.1 | 4 | 531 | |||
XM_006234040.1 | XP_006234102.1 | 5 | 651 | |||
GenBank mRNA | BC079472.1 | - | 4 | 633 | 7122783 | |
FQ223877.1 | - | 5 | 656 |
