siRNA Details
Gene Symbol: Rpl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212511.2 | NP_997676.1 | 2, 3 | 175 | ||
GenBank mRNA | BC058499.1 | - | 2, 3 | 179 | 6919623 | |
FQ211496.1 | - | 2, 3 | 175 | |||
FQ212202.1 | - | 2, 3 | 175 | |||
FQ213515.1 | - | 2, 3 | 175 | |||
FQ217126.1 | - | 2, 3 | 201 | |||
FQ221404.1 | - | 2, 3 | 175 | |||
FQ222970.1 | - | 2, 3 | 175 | |||
FQ222977.1 | - | 2, 3 | 175 | |||
FQ223272.1 | - | 2, 3 | 181 | |||
FQ223441.1 | - | 2, 3 | 175 | |||
FQ224793.1 | - | 2, 3 | 154 | |||
FQ224804.1 | - | 2, 3 | 175 | |||
FQ230871.1 | - | 2, 3 | 175 | |||
X51705.1 | - | 2, 3 | 183 |

More Information
Other Species Targeted

Gene Ontology Categories:
