siRNA Details
Gene Symbol: Arpc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037767.2 | NP_001032856.2 | 3 | 397 | ||
GenBank mRNA | BC098820.1 | - | 2 | 538 | ||
BC127445.1 | - | 3 | 326 | |||
FM029541.1 | - | 3 | 397 |

More Information
Other Species Targeted

Gene Ontology Categories:
