siRNA Details
Gene Symbol: Arpc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037767.2 | NP_001032856.2 | 3 | 378 | ||
GenBank mRNA | BC098820.1 | - | 2 | 519 | ||
BC127445.1 | - | 3 | 307 | |||
FM029541.1 | - | 3 | 378 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
