siRNA Details
Gene Symbol: Fastk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235892.2 | XP_006235954.1 | 2 | 527 | ||
XM_006235894.2 | XP_006235956.1 | 2 | 514 | |||
XM_006235895.1 | XP_006235957.1 | 2 | 511 | |||
XM_006235896.2 | XP_006235958.1 | 2 | 528 | |||
GenBank mRNA | BC083715.1 | - | 2 | 531 | 7190690 | |
FQ215763.1 | - | 3 | 389 | |||
FQ228972.1 | - | 2 | 450 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023229, AK152369, AK151859, BC013547, AK002552, and others.
Gene Ontology Categories:
