siRNA Details
Gene Symbol: Srpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106575.2 | NP_001100045.1 | 3 | 435 | ||
XM_006235903.2 | XP_006235965.1 | 3 | 3431 | |||
XM_008762635.1 | XP_008760857.1 | 3 | 2853 | |||
XM_008762636.1 | XP_008760858.1 | 3 | 3431 | |||
XM_008762637.1 | XP_008760859.1 | 3 | 3430 | |||
GenBank mRNA | BC161879.1 | - | 3 | 435 | 7125533 | |
FQ234485.1 | - | 3 | 448 |
