siRNA Details
Gene Symbol: Phtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106577.1 | NP_001100047.1 | 7, 8 | 818 | ||
XM_008762640.1 | XP_008760862.1 | 7, 8 | 773 | |||
XM_008762641.1 | XP_008760863.1 | 6, 7 | 2356 | |||
XM_008762642.1 | XP_008760864.1 | 7, 8 | 2368 | |||
XM_008762643.1 | XP_008760865.1 | 6, 7 | 670 | |||
XM_008762644.1 | XP_008760866.1 | 7, 8 | 2368 |
