siRNA Details
Gene Symbol: Las1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170590.1 | NP_001164061.1 | 8, 9 | 1199 | ||
XM_008773298.1 | XP_008771520.1 | 8, 9 | 1190 | |||
GenBank mRNA | BC169024.1 | - | 1, 2 | 712 | 9029735 | |
CB751131.1 | - | 3, 4 | 379 | |||
CB811034.1 | - | 1, 2 | 256 |
