siRNA Details
Gene Symbol: Lsm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106585.1 | NP_001100055.1 | 2, 3 | 216 | ||
GenBank mRNA | BC167094.1 | - | 2, 3 | 87 | 7462085 | |
FQ211862.1 | - | 182 | ||||
FQ212479.1 | - | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
