siRNA Details
Gene Symbol: Chchd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106588.1 | NP_001100058.1 | 5 | 562 | ||
GenBank mRNA | FQ216356.1 | - | 5 | 545 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chchd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106588.1 | NP_001100058.1 | 5 | 562 | ||
GenBank mRNA | FQ216356.1 | - | 5 | 545 |