siRNA Details
Gene Symbol: Akr1b10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013084.1 | NP_001013102.1 | 8 | 868 | ||
GenBank mRNA | BC079133.1 | - | 8 | 868 | 7127413 | |
FQ215899.1 | - | 756 |
Gene Symbol: LOC100910708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762861.1 | XP_008761083.1 | 7 | 1016 |
