siRNA Details
Gene Symbol: Mrps33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047863.1 | NP_001041328.1 | 1 | 163 | ||
XM_006236305.2 | XP_006236367.1 | 2 | 276 | |||
GenBank mRNA | DQ480753.1 | - | 1 | 163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
