siRNA Details
Gene Symbol: Rarres2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013427.1 | NP_001013445.1 | 5 | 484 | ||
XM_006236415.2 | XP_006236477.1 | 5 | 511 | |||
XM_006236416.1 | XP_006236478.1 | 5 | 569 | |||
GenBank mRNA | BC091177.1 | - | 5 | 484 | 7323591 | |
FQ209660.1 | - | 4 | 678 | |||
FQ209771.1 | - | 5 | 498 | |||
FQ210577.1 | - | 5 | 499 | |||
FQ210699.1 | - | 5 | 499 | |||
FQ228347.1 | - | 5 | 533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
