siRNA Details
Gene Symbol: Rarres2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013427.1 | NP_001013445.1 | 3 | 352 | ||
XM_006236415.2 | XP_006236477.1 | 3 | 379 | |||
XM_006236416.1 | XP_006236478.1 | 3 | 440 | |||
GenBank mRNA | BC091177.1 | - | 3 | 352 | 7323591 | |
FQ209771.1 | - | 3 | 366 | |||
FQ210577.1 | - | 3 | 367 | |||
FQ210699.1 | - | 3 | 367 | |||
FQ228347.1 | - | 3 | 401 |
