siRNA Details
Gene Symbol: Gimap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011968.3 | NP_001011968.1 | 1 | 202 | ||
GenBank mRNA | AJ633683.1 | - | 1 | 205 | ||
BC089859.1 | - | 111 | 7384582 | |||
DQ125342.1 | - | 1 | 112 | |||
DQ125343.1 | - | 112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
