siRNA Details
Gene Symbol: Tmem176a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039008.1 | NP_001034097.1 | 5, 6 | 703 | ||
GenBank mRNA | BC098714.1 | - | 5, 6 | 703 | 7323021 | |
FQ209561.1 | - | 6, 7 | 743 | |||
FQ209602.1 | - | 6, 7 | 754 | |||
FQ210479.1 | - | 6, 7 | 800 | |||
FQ210827.1 | - | 6, 7 | 754 | |||
FQ219180.1 | - | 6, 7 | 743 | |||
FQ231468.1 | - | 6, 7 | 749 |
