siRNA Details
Gene Symbol: Mad2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106594.1 | NP_001100064.1 | 5 | 672 | ||
XM_006236619.2 | XP_006236681.1 | 5 | 762 | |||
XM_006236620.2 | XP_006236682.1 | 5 | 762 | |||
XM_006236621.2 | XP_006236683.1 | 5 | 762 | |||
GenBank mRNA | FQ222936.1 | - | 5 | 648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
