siRNA Details
Gene Symbol: Usp39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106597.1 | NP_001100067.1 | 10 | 1372 | ||
XM_006236659.2 | XP_006236721.1 | 11 | 2401 | |||
GenBank mRNA | BC166516.1 | - | 1830 | 7131845 |
