siRNA Details
Gene Symbol: Rnf181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007647.1 | NP_001007648.1 | 5 | 816 | ||
GenBank mRNA | BC079313.1 | - | 5 | 816 | 7134895 | |
FQ213488.1 | - | 4 | 465 | |||
FQ230795.1 | - | 4 | 415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
