siRNA Details
Gene Symbol: Rtkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_184046.1 | NP_908935.1 | 1997 | |||
XM_006236729.2 | XP_006236791.1 | 4025 | ||||
XM_006236731.2 | XP_006236793.1 | 3815 | ||||
XM_008763054.1 | XP_008761276.1 | 1952 | ||||
XM_008763055.1 | XP_008761277.1 | 1789 | ||||
GenBank mRNA | AY348867.1 | - | 1997 | |||
AY348868.1 | - | 1295 |
Gene Symbol: LOC103692165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763034.1 | XP_008761256.1 | 12 | 4025 | ||
XM_008763035.1 | XP_008761257.1 | 12 | 1952 | |||
XM_008763036.1 | XP_008761258.1 | 12 | 1789 | |||
XM_008763037.1 | XP_008761259.1 | 11 | 3815 |
