siRNA Details
Gene Symbol: Nfu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106606.2 | NP_001100076.2 | 5 | 447 | ||
XM_008763063.1 | XP_008761285.1 | 5 | 680 | |||
GenBank mRNA | FM058785.1 | - | 5 | 447 | ||
FM064033.1 | - | 4 | 273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
