siRNA Details
Gene Symbol: Nfu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106606.2 | NP_001100076.2 | 5 | 393 | ||
XM_008763063.1 | XP_008761285.1 | 5 | 626 | |||
GenBank mRNA | FM058785.1 | - | 5 | 393 | ||
FM064033.1 | - | 4 | 219 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001170591, NM_020045, AY335195, BC018355, AJ132616, and others.
Gene Ontology Categories:
