siRNA Details
Gene Symbol: Gfpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005879.1 | NP_001005879.1 | 18 | 1973 | ||
XM_006236823.2 | XP_006236885.1 | 19 | 2099 | |||
GenBank mRNA | BC083889.1 | - | 18 | 1973 | 7132442 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013528, AK019852, U00932, BC002283, BC050762, and others.
Gene Ontology Categories:
