siRNA Details
Gene Symbol: Gkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039686.1 | NP_001034775.1 | 3 | 224 | ||
XM_008763064.1 | XP_008761286.1 | 2 | 1752 | |||
XM_008763065.1 | XP_008761287.1 | 6 | 708 | |||
GenBank mRNA | AY943907.1 | - | 3 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
