siRNA Details
Gene Symbol: V1ra14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008898.1 | NP_001008898.1 | 1 | 875 | ||
GenBank mRNA | AY510301.1 | - | 1 | 875 |
Gene Symbol: Vom1r103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167553.1 | NP_001161025.1 | 1 | 845 |
Gene Symbol: Vom1r105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173113.1 | NP_775136.1 | 2 | 1069 | ||
GenBank mRNA | U36785.1 | - | 2 | 1069 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
