siRNA Details
Gene Symbol: Ccdc174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009659.2 | NP_001009659.2 | 5 | 434 | ||
XM_006236907.2 | XP_006236969.1 | 5 | 471 | |||
GenBank mRNA | BC087050.1 | - | 5 | 434 | 7114831 |

More Information
Other Species Targeted

