siRNA Details
Gene Symbol: Eif4e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106612.1 | NP_001100082.1 | 6 | 514 | ||
XM_008763159.1 | XP_008761381.1 | 6 | 928 | |||
XM_008763160.1 | XP_008761382.1 | 8 | 998 | |||
XM_008763161.1 | XP_008761383.1 | 7 | 862 | |||
GenBank mRNA | BC168936.1 | - | 6 | 501 | 7124999 |
