siRNA Details
Gene Symbol: Shq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134713.1 | NP_001128185.1 | 903 | |||
XM_008763162.1 | XP_008761384.1 | 911 | ||||
GenBank mRNA | BC168172.1 | - | 903 | 9026449 |
