siRNA Details
Gene Symbol: Nampt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177928.3 | NP_808789.1 | 1, 2 | 89 | ||
GenBank mRNA | AB081730.1 | - | 1, 2 | 64 | ||
AY831728.1 | - | 1, 2 | 62 | |||
BC085681.1 | - | 1, 2 | 89 | 7192109 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_021524, BY068469, AK153926, AK166270, AY679720, and others.
Gene Ontology Categories:
