siRNA Details
Gene Symbol: Setd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106614.1 | NP_001100084.1 | 20 | 3847 | ||
XM_006237012.2 | XP_006237074.1 | 19 | 3514 | |||
XM_006237013.2 | XP_006237075.1 | 17 | 7633 | |||
XM_006237014.2 | XP_006237076.1 | 18 | 7615 | |||
XM_006237015.1 | XP_006237077.1 | 19 | 3276 | |||
XM_006237016.2 | XP_006237078.1 | 18 | 3475 | |||
XM_006237017.2 | XP_006237079.1 | 17 | 7576 | |||
XM_006237018.2 | XP_006237080.1 | 18 | 3418 | |||
XM_006237020.2 | XP_006237082.1 | 18 | 4538 | |||
XM_006237021.2 | XP_006237083.1 | 19 | 3459 | |||
XM_008763184.1 | XP_008761406.1 | 18 | 7672 | |||
XM_008763185.1 | XP_008761407.1 | 19 | 6873 | |||
XM_008763186.1 | XP_008761408.1 | 19 | 4364 | |||
XM_008763187.1 | XP_008761409.1 | 18 | 4402 | |||
XM_008763188.1 | XP_008761410.1 | 19 | 4308 | |||
XM_008763189.1 | XP_008761411.1 | 16 | 3068 | |||
GenBank mRNA | FQ223178.1 | - | 19 | 3304 |

More Information
Other Species Targeted

