siRNA Details
Gene Symbol: Setd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106614.1 | NP_001100084.1 | 9 | 1455 | ||
XM_006237012.2 | XP_006237074.1 | 8 | 1122 | |||
XM_006237013.2 | XP_006237075.1 | 6 | 5241 | |||
XM_006237014.2 | XP_006237076.1 | 7 | 5223 | |||
XM_006237015.1 | XP_006237077.1 | 8 | 884 | |||
XM_006237016.2 | XP_006237078.1 | 7 | 1083 | |||
XM_006237017.2 | XP_006237079.1 | 6 | 5184 | |||
XM_006237018.2 | XP_006237080.1 | 7 | 1026 | |||
XM_006237020.2 | XP_006237082.1 | 7 | 2146 | |||
XM_006237021.2 | XP_006237083.1 | 8 | 1067 | |||
XM_008763184.1 | XP_008761406.1 | 7 | 5280 | |||
XM_008763185.1 | XP_008761407.1 | 8 | 4481 | |||
XM_008763186.1 | XP_008761408.1 | 8 | 1972 | |||
XM_008763187.1 | XP_008761409.1 | 7 | 2010 | |||
XM_008763188.1 | XP_008761410.1 | 8 | 1916 | |||
XM_008763189.1 | XP_008761411.1 | 5 | 676 | |||
GenBank mRNA | FQ223178.1 | - | 8 | 912 |

More Information
Other Species Targeted

