siRNA Details
Gene Symbol: Setd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106614.1 | NP_001100084.1 | 11 | 1873 | ||
XM_006237012.2 | XP_006237074.1 | 10 | 1540 | |||
XM_006237013.2 | XP_006237075.1 | 8 | 5659 | |||
XM_006237014.2 | XP_006237076.1 | 9 | 5641 | |||
XM_006237015.1 | XP_006237077.1 | 10 | 1302 | |||
XM_006237016.2 | XP_006237078.1 | 9 | 1501 | |||
XM_006237017.2 | XP_006237079.1 | 8 | 5602 | |||
XM_006237018.2 | XP_006237080.1 | 9 | 1444 | |||
XM_006237020.2 | XP_006237082.1 | 9 | 2564 | |||
XM_006237021.2 | XP_006237083.1 | 10 | 1485 | |||
XM_008763184.1 | XP_008761406.1 | 9 | 5698 | |||
XM_008763185.1 | XP_008761407.1 | 10 | 4899 | |||
XM_008763186.1 | XP_008761408.1 | 10 | 2390 | |||
XM_008763187.1 | XP_008761409.1 | 9 | 2428 | |||
XM_008763188.1 | XP_008761410.1 | 10 | 2334 | |||
XM_008763189.1 | XP_008761411.1 | 7 | 1094 | |||
GenBank mRNA | FQ223178.1 | - | 10 | 1330 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
