siRNA Details
Gene Symbol: Arpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106615.2 | NP_001100085.2 | 3 | 193 | ||
GenBank mRNA | BC167049.1 | - | 3 | 181 | 7442696 | |
CK478610.1 | - | 3 | 193 | |||
FQ228740.1 | - | 3 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
