siRNA Details
Gene Symbol: Il17rc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170565.1 | NP_001164036.1 | 9 | 959 | ||
GenBank mRNA | CB793227.1 | - | 4 | 310 | ||
CB802860.1 | - | 4 | 222 |

More Information
Other Species Targeted

Gene Ontology Categories:
