siRNA Details
Gene Symbol: Csgalnact2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106616.1 | NP_001100086.1 | 3 | 1183 | ||
XM_006237181.2 | XP_006237243.1 | 3 | 956 | |||
XM_006237182.2 | XP_006237244.1 | 3 | 1197 | |||
GenBank mRNA | FQ221326.1 | - | 3 | 1095 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
