siRNA Details
Gene Symbol: Atp6v1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198745.2 | NP_942040.1 | 3 | 261 | ||
GenBank mRNA | BC059155.1 | - | 3 | 265 | 6887702 | |
FQ212928.1 | - | 3 | 261 | |||
FQ212966.1 | - | 3 | 256 | |||
FQ213001.1 | - | 3 | 261 | |||
FQ213103.1 | - | 3 | 258 | |||
FQ213345.1 | - | 3 | 263 | |||
FQ213479.1 | - | 3 | 262 | |||
FQ213531.1 | - | 3 | 263 | |||
FQ213667.1 | - | 3 | 279 | |||
FQ213669.1 | - | 3 | 263 | |||
FQ213903.1 | - | 3 | 257 | |||
FQ214240.1 | - | 3 | 257 | |||
FQ214254.1 | - | 3 | 263 | |||
FQ214381.1 | - | 3 | 261 | |||
FQ214406.1 | - | 3 | 256 | |||
FQ214476.1 | - | 3 | 266 | |||
FQ214497.1 | - | 3 | 229 | |||
FQ214920.1 | - | 3 | 261 | |||
FQ216629.1 | - | 3 | 261 | |||
FQ224865.1 | - | 3 | 262 | |||
FQ226034.1 | - | 3 | 256 | |||
FQ230006.1 | - | 3 | 261 | |||
FQ230733.1 | - | 3 | 256 | |||
FQ233079.1 | - | 3 | 261 | |||
FQ233569.1 | - | 3 | 263 |
