siRNA Details
Gene Symbol: Atp6v1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198745.2 | NP_942040.1 | 2 | 160 | ||
GenBank mRNA | BC059155.1 | - | 2 | 164 | 6887702 | |
FQ212928.1 | - | 2 | 160 | |||
FQ212966.1 | - | 2 | 155 | |||
FQ213001.1 | - | 2 | 160 | |||
FQ213103.1 | - | 2 | 157 | |||
FQ213345.1 | - | 2 | 162 | |||
FQ213479.1 | - | 2 | 161 | |||
FQ213531.1 | - | 2 | 162 | |||
FQ213667.1 | - | 2 | 178 | |||
FQ213669.1 | - | 2 | 162 | |||
FQ213903.1 | - | 2 | 156 | |||
FQ214240.1 | - | 2 | 156 | |||
FQ214254.1 | - | 2 | 162 | |||
FQ214381.1 | - | 2 | 160 | |||
FQ214406.1 | - | 2 | 155 | |||
FQ214440.1 | - | 2 | 162 | |||
FQ214476.1 | - | 2 | 165 | |||
FQ214497.1 | - | 2 | 128 | |||
FQ214920.1 | - | 2 | 160 | |||
FQ224865.1 | - | 2 | 161 | |||
FQ226034.1 | - | 2 | 155 | |||
FQ230006.1 | - | 2 | 160 | |||
FQ230733.1 | - | 2 | 155 | |||
FQ233079.1 | - | 2 | 160 | |||
FQ233569.1 | - | 2 | 162 |
