siRNA Details
Gene Symbol: A1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037975.2 | NP_001033064.2 | 17 | 2100 | ||
XM_006237281.2 | XP_006237343.1 | 17 | 2112 | |||
GenBank mRNA | J03552.1 | - | 17 | 2100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
