siRNA Details
Gene Symbol: Pex26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106618.2 | NP_001100088.1 | 2, 3 | 683 | ||
XM_006237235.2 | XP_006237297.1 | 2, 3 | 1309 | |||
GenBank mRNA | BC166463.1 | - | 2, 3 | 683 | 9030430 |
