siRNA Details
Gene Symbol: Lag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212513.2 | NP_997678.2 | 6 | 1203 | ||
XM_006237348.2 | XP_006237410.1 | 6 | 1428 | |||
XM_008763282.1 | XP_008761504.1 | 7 | 1309 | |||
XM_008763283.1 | XP_008761505.1 | 5 | 953 | |||
GenBank mRNA | AY230414.1 | - | 6 | 1093 | ||
BC091201.1 | - | 6 | 1203 | 7380279 | ||
DQ438937.1 | - | 6 | 1081 |
