siRNA Details
Gene Symbol: Ing4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079887.1 | NP_001073356.1 | 6 | 583 | ||
XM_006237349.2 | XP_006237411.1 | 6 | 694 | |||
XM_006237350.2 | XP_006237412.1 | 6 | 691 | |||
GenBank mRNA | BC086570.1 | - | 5 | 789 | ||
BC097942.1 | - | 4 | 972 | |||
BC127511.1 | - | 6 | 583 | 7376723 | ||
FQ213328.1 | - | 3 | 220 | |||
FQ232370.1 | - | 2 | 217 |

More Information
Other Species Targeted

Gene Ontology Categories:
