siRNA Details
Gene Symbol: Ing4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079887.1 | NP_001073356.1 | 2 | 99 | ||
XM_006237349.2 | XP_006237411.1 | 2 | 198 | |||
XM_006237350.2 | XP_006237412.1 | 2 | 198 | |||
GenBank mRNA | BC086570.1 | - | 2 | 99 | ||
BC097942.1 | - | 2 | 100 | |||
BC127511.1 | - | 2 | 99 | 7376723 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
