siRNA Details
Gene Symbol: Wbp11l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763449.1 | XP_008761671.1 | 395 | |||
XM_008763450.1 | XP_008761672.1 | 397 |
Gene Symbol: Wbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763375.1 | 5 | 395 | |||
GenBank mRNA | BC087082.1 | - | 5 | 308 | 7189215 |
