siRNA Details
Gene Symbol: Strap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011969.1 | NP_001011969.1 | 6 | 807 | ||
GenBank mRNA | BC083714.1 | - | 6 | 807 | 7190676 |
Gene Symbol: LOC100910575 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749864.3 | XP_003749912.2 | 6 | 828 |
