siRNA Details
Gene Symbol: Mrps35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106628.1 | NP_001100098.1 | 5, 6 | 538 | ||
XM_006237676.1 | XP_006237738.1 | 5, 6 | 543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
