siRNA Details
Gene Symbol: Jph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106630.1 | NP_001100100.1 | 5 | 2129 | ||
XM_006237719.2 | XP_006237781.1 | 5 | 2129 | |||
XM_006237720.2 | XP_006237782.1 | 5 | 2129 | |||
XM_006237721.2 | XP_006237783.1 | 5 | 2129 | |||
XM_006237722.2 | XP_006237784.1 | 5 | 2129 | |||
XM_006237723.2 | XP_006237785.1 | 5 | 2129 | |||
XM_008763475.1 | XP_008761697.1 | 5 | 2129 |
