siRNA Details
Gene Symbol: Mybl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106632.1 | NP_001100102.1 | 15 | 2444 | ||
XM_006237749.2 | XP_006237811.1 | 15 | 2555 | |||
XM_006237750.2 | XP_006237812.1 | 15 | 3365 | |||
XM_006237751.2 | XP_006237813.1 | 14 | 2450 |
