siRNA Details
Gene Symbol: Mrpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237805.2 | XP_006237867.1 | 5 | 694 | ||
GenBank mRNA | FQ214529.1 | - | 5 | 656 | ||
FQ216110.1 | - | 5 | 687 | |||
FQ216137.1 | - | 5 | 635 | |||
FQ221067.1 | - | 5 | 654 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
