siRNA Details
Gene Symbol: Mrpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237805.2 | XP_006237867.1 | 5 | 896 | ||
GenBank mRNA | FQ214529.1 | - | 5 | 858 | ||
FQ216110.1 | - | 5 | 889 | |||
FQ216137.1 | - | 5 | 837 |

More Information
Other Species Targeted

Gene Ontology Categories:
