siRNA Details
Gene Symbol: Car8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009662.1 | NP_001009662.1 | 7 | 796 | ||
GenBank mRNA | BC087586.1 | - | 7 | 796 | 7318288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
